Merck
The development of potential new fluorine-18 labelled radiotracers for imaging the GABA(A) receptor.
Bioorganic & medicinal chemistry letters (2012-12-26)
Alexander Jackson, Benedicte B Guilbert, Stuart D Plant, Julian Goggi, Mark R Battle, John L Woodcraft, Alessandra Gaeta, Clare L Jones, Denis R Bouvet, Paul A Jones, Dennis M O'Shea, Penny Hao Zheng, Samantha L Brown, Amanda L Ewan, William Trigg
ABSTRAKT
Positron emission tomography (PET) using the tracer [(11)C]Flumazenil has shown changes in the distribution and expression of the GABA(A) receptor in a range of neurological conditions and injury states. We aim to develop a fluorine-18 labelled PET agent with comparable properties to [(11)C]Flumazenil. In this study we make a direct comparison between the currently known fluorine-18 labelled GABA(A) radiotracers and novel imidazobenzodiazepine ligands. A focussed library of novel compound was designed and synthesised where the fluorine containing moiety and the position of attachment is varied. The in vitro affinity of twenty-two compounds for the GABA(A) receptor was measured. Compounds containing a fluoroalkyl amide or a longer chain ester group were eliminated due to low potency. The fluorine-18 radiochemistry of one compound from each structural type was assessed to confirm that an automated radiosynthesis in good yield was feasible. Eleven of the novel compounds assessed appeared suitable for in vivo assessment as PET tracers.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Flumazenil, >99% (HPLC), solid