Merck
Abstracts of the Sixth International Congress of the GRS and the IGF Society. Munich, Germany. October 17-20, 2012.
Growth hormone & IGF research : official journal of the Growth Hormone Research Society and the International IGF Research Society (2013-02-21)