Merck
  • Application of reduced graphene oxide and carbon nanotube modified electrodes for measuring the enzymatic activity of alcohol dehydrogenase.
Application of reduced graphene oxide and carbon nanotube modified electrodes for measuring the enzymatic activity of alcohol dehydrogenase.
Food chemistry (2013-03-19)
Xianlong Wang, Li Li, Yanping Wang, Chongzheng Xu, Bo Zhao, Xiaodi Yang
ABSTRAKT
An electrochemical method was developed to measure the enzymatic activity of alcohol dehydrogenase (ADH) by monitoring the amount of reduced nicotinamide adenine dinucleotide (NADH) generated in the catalysed oxidation of ethanol by ADH. The concentration of NADH was determined by amperometric measurements, which recorded the oxidation current of NADH versus time on reduced graphene oxide and functionalised multi-walled carbon nanotube modified electrodes. The initial reaction rates and the apparent Michaelis constants of the enzymatic reaction were obtained in the absence and presence of Al(3+) and nanometre-sized tridecameric aluminium polycationic (nano-Al(13)) species. The results showed that Al(3+) and nano-Al(13) exhibited inhibitory effect on the enzymatic activity of ADH. Fluorescence and circular dichroism spectra indicated the inhibitory effect was likely caused by the conformational changes of ADH and/or NADH induced by Al(3+) and nano-Al(13).
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Aluminum, powder, ≥91% (complexometric)
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, ReagentPlus®, beads, 5-15 mm, 99.9% trace metals basis
Sigma-Aldrich
Aluminum, ACS reagent, 99%, wire, wire diam. ~1.5 mm
Sigma-Aldrich
Aluminum, foil, thickness 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.13 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 0.45-0.55 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-12 mm, 99.99% trace metals basis
Sigma-Aldrich
Aluminum, foil, thickness 1.0 mm, 99.999% trace metals basis
Aluminum, IRMM®, certified reference material, 0.1 mm foil