- Concentrations of thiabendazole and parasite-specific IgG antibodies in the cerebrospinal fluid of a patient with disseminated strongyloidiasis.
Concentrations of thiabendazole and parasite-specific IgG antibodies in the cerebrospinal fluid of a patient with disseminated strongyloidiasis.
The Journal of infectious diseases (1987-09-01)
J C Arroyo, A Brown
PMID3611836
MATERIAŁY