Merck
[Recent development of C-terminal sequence analysis of proteins].
Tanpakushitsu kakusan koso. Protein, nucleic acid, enzyme (1998-06-23)
A Tsugita
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Sodium pentafluoropropionate, 98%
Sigma-Aldrich
Pentafluoropropionic acid, 97%