Merck
Site-specific derivatization of RNA with photocrosslinkable groups.
Methods (San Diego, Calif.) (1999-04-20)
M M Konarska
ABSTRAKT
Derivatization of RNA with heterobifunctional photocrosslinking reagents becomes an increasingly popular method for the analysis of structural properties of ribonucleoprotein complexes. This article describes a simple chemical modification-derivatization strategy used to introduce selected chemical groups at specific internal positions within the RNA ribose backbone. The strategy is based on the coupling of a haloacetyl adduct to a thiol residue in the phosphodiester bond. The use of a number of RNA probes derivatized with several different photoreactive groups can provide invaluable information on the structural distribution of components in complex ribonucleoprotein assemblies.