Merck
The neuropeptide FF analogue, 1DMe, reduces in vivo dynorphin release from the rat spinal cord.
Journal of neurochemistry (2002-06-18)
Sébastien Ballet, Joao Braz, Annie Mauborgne, Sylvie Bourgoin, Jean-Marie Zajac, Michel Hamon, François Cesselin
ABSTRAKT
Intrathecal infusion of the neuropeptide FF analogue, [D-Tyr1, (NMe)Phe3]neuropeptide FF (1DMe; 0.1 microm-0.1 mm) in anaesthetized rats produced a concentration-dependent decrease in the spinal outflow of dynorphin A (1-8)-like material, which persisted for at least 90 min after treatment with 10 microm-0.1 mm of the compound. Co-administration of d-Phe-Cys-Tyr-D-Trp-Orn-Thr-Pen-Thr-NH2 (CTOP; 1 microm) to block spinal micro-opioid receptors did not modify this effect, whereas naltrindole (10 microm) totally prevented it and nor-binaltorphimine (10 microm) reduced the post-effect. These data suggest that 1DMe triggers the release of endogenous opioids that stimulate mainly delta-opioid receptors, and secondarily kappa-opioid receptors, thereby exerting a negative influence on dynorphin A (1-8)-like material outflow. Because dynorphin has pronociceptive properties, such a decrease in spinal dynorphin A (1-8)-like material release might underlie the long-lasting antinociceptive effects of intrathecally administered neuropeptide FF and analogues.