- Development of a yeast assay for rapid screening of inhibitors of human-derived Pneumocystis carinii dihydrofolate reductase.
Development of a yeast assay for rapid screening of inhibitors of human-derived Pneumocystis carinii dihydrofolate reductase.
Antimicrobial agents and chemotherapy (2002-08-17)
Liang Ma, Qiuyao Jia, Joseph A Kovacs
PMID12183283
ABSTRAKT
Human-derived Pneumocystis carinii dihydrofolate reductase (DHFR) was expressed in a Saccharomyces cerevisiae strain whose growth depends on complementation by this enzyme. We utilized a quantitative assay to measure the sensitivity of this yeast strain to DHFR inhibitors. This assay should be useful for identifying new inhibitors of human-derived P. carinii DHFR.