- beta-peptides as catalysts: poly-beta-leucine as a catalyst for the Juliá-Colonna asymmetric epoxidation of enones.
beta-peptides as catalysts: poly-beta-leucine as a catalyst for the Juliá-Colonna asymmetric epoxidation of enones.
Chemical communications (Cambridge, England) (2002-09-21)
P E Coffey, K H Drauz, S M Roberts, J Skidmore, J A Smith
PMID12240060
ABSTRAKT
Poly-beta-leucines have been evaluated as catalysts for the Juliá-Colonna asymmetric epoxidation of enones; the beta 3-isomer was found to be an effective catalyst for the epoxidation of chalcone (70% ee) and some analogues.