Merck
Photoinductive efficiency of soil extracted humic and fulvic acids.
Chemosphere (2002-10-05)
J P Aguer, C Richard, O Trubetskaya, O Trubetskoj, J Lévèque, F Andreux
ABSTRAKT
Humic and fulvic acids extracted from soils of different genesis were investigated for their ability to photoinduce the transformation of fenuron (2 x 10(-4) mol(-1)) at 365 nm. The ratio of the initial rate of fenuron consumption over the rate of light absorption by humic substances was found to be higher for fulvic acids (range 2.0 x 10(-3) to 9.0 x 10(-5)) than for humic acids (range 1.7 x 10(-4) to - 3.6 x 10(-5)). Within the FAs population, this ratio decreased as the specific absorption coefficient at 365 nm increased. It seems therefore that most of 365-nm absorbing components have no photoinductive activity and even reduce that of photoinductive chromophores.