Merck
Musical hallucinations during a treatment with benzodiazepine.
Canadian journal of psychiatry. Revue canadienne de psychiatrie (2002-11-08)
François Curtin, Charles Redmund
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Lormetazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®