Merck
  • [The biosynthesis of low-molecular-weight nitrogen-containing secondary metabolite-alkaloids by the resident strains of Penicillium chrysogenum and Penicillium expansum isolated on the board of the Mir space station ].
[The biosynthesis of low-molecular-weight nitrogen-containing secondary metabolite-alkaloids by the resident strains of Penicillium chrysogenum and Penicillium expansum isolated on the board of the Mir space station ].
Mikrobiologiia (2003-01-16)
A G Kozlovskiĭ, V P Zhelifonova, V M Adanin, T V Antipova, A V Shnyreva, A N Viktorov
ABSTRAKT
The analysis of the absorption spectra of the low-molecular-weight nitrogen-containing secondary metabolites--alkaloids--of 4 Penicillium chrysogenum strains and 6 Penicillium expansum strains isolated on board the Mir space station showed that all these strains synthesize metabolites of alkaloid origin (roquefortine, 3,12-dihydroroquefortine, meleagrin, viridicatin, viridicatol, isorugulosuvin, rugulosuvin B, N-acetyl-tryptamine, and a "yellow metabolite" containing the benzoquinone chromophore).