- Isolation of a new dual-functional caffeine synthase gene encoding an enzyme for the conversion of 7-methylxanthine to caffeine from coffee (Coffea arabica L.).
Isolation of a new dual-functional caffeine synthase gene encoding an enzyme for the conversion of 7-methylxanthine to caffeine from coffee (Coffea arabica L.).
FEBS letters (2003-01-16)
Kouichi Mizuno, Akira Okuda, Misako Kato, Naho Yoneyama, Hiromi Tanaka, Hiroshi Ashihara, Tatsuhito Fujimura
PMID12527364
ABSTRAKT
In coffee and tea plants, caffeine is synthesized from xanthosine via a pathway that includes three methylation steps. We report the isolation of a bifunctional coffee caffeine synthase (CCS1) clone from coffee endosperm by reverse transcription-polymerase chain reaction (RT-PCR) and rapid amplification of cDNA ends (RACE) technique using previously reported sequence information for theobromine synthases (CTSs). The predicted amino acid sequences of CCS1 are more than 80% identical to CTSs and are about 40% similar to those of tea caffeine synthase (TCS1). Interestingly, CCS1 has dual methylation activity like tea TCS1.