Merck
A membrane potential-sensitive dye for vascular smooth muscle cells assays.
Biochemical and biophysical research communications (2003-01-22)
Fabiana S Sguilla, Antonio C Tedesco, Lusiane M Bendhack
ABSTRAKT
Changes in membrane potential of rat aorta smooth muscle cells were investigated using the bis-oxonol sensitive probe DIBAC2(3). We compared the changes in membrane potential induced by a high external KCl concentration in aorta smooth muscle cells from normotensive 2 kidney (2K) and from renal hypertensive 2 kidney-1 clip (2K-1C) rats. The spectral properties of the membrane potential were first characterized in aqueous buffers and in cultured smooth muscle cells from 2K and 2K-1C rat aortas. Fluorescence emission and the images were recorded using a laser scanning confocal microscope. The relationship between fluorescence intensity (FI) and membrane potential (psi(m)) as a function of the increasing extracellular KCl concentration was linear in the 5-40 mmol/L KCl range in both 2K and 2K-1C rat aorta cells. Cell membranes from 2K-1C rat aorta cells were more depolarized (-55 mV) than 2K rat aorta cells (-65 mV). The results show that in 2K-1C aorta cells only 10 mmol/L KCl was needed to induce complete membrane depolarization while in 2K cells 40 mmol/L KCl was needed to induce a similar effect. This study clearly shows that the method is suitable to measure the membrane potential in cultured smooth muscle cells.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
DISBAC2(3), Voltage Sensitive Probe, ≥98% (HPLC)