Merck
Dietary low-dose sucrose modulation of IQ-induced genotoxicity in the colon and liver of Big Blue rats.
Mutation research (2003-06-06)
Peter Møller, Max Hansen, Herman Autrup, Jette Bornholt, Ulla Vogel, Anne-Marie Mølck, Håkan Wallin, Lars O Dragsted, Lotte Risom, Henrik E Poulsen, Steffen Loft
ABSTRAKT
Earlier studies have indicated that sucrose increases 2-amino-3-methylimidazo[4,5-f]quinoline (IQ)-induced aberrant crypt foci in the colon. In this study, we investigated the role of sucrose in IQ-induced genotoxicity of the colon mucosa and liver. Big Blue rats were fed with IQ (20 ppm in feed) and/or sucrose (3.45 or 6.85 wt.% in feed) for 3 weeks. IQ increased DNA strand breaks in the colon, whereas the mutation frequency was increased in the liver. The level of IQ-induced DNA adducts was elevated in both colon mucosa cells and liver. In the liver, high sucrose intake increased the level of DNA adducts above that of IQ and low sucrose intake. Oxidative DNA damage detected in terms of 7-hydro-8-oxo-2'-deoxyguanosine by HPLC-EC, or endonuclease III or formamidopyrimidine DNA glycosylase sensitive sites were unaltered in the colon and liver. Expression of ERCC1 and OGG1 mRNA levels were unaffected by IQ or sucrose feeding. Biomarkers of oxidative stress, including Vitamin C, malondialdehyde and protein oxidations (gamma-glutamyl semialdehyde and 2-amino adipic semialdehyde) were unaltered in plasma and in liver. In conclusion, sucrose feeding increases IQ-induced genotoxicity in liver but not in colon, suggesting different mechanisms for sucrose and IQ in colon mutagenesis.