- [The principles of left-right hemispheric interactions of pharmacological preparations exemplified by Brinaldix].
[The principles of left-right hemispheric interactions of pharmacological preparations exemplified by Brinaldix].
Izvestiia Akademii nauk. Seriia biologicheskaia (1992-05-01)
L A Piruzian, L L Klimenko
PMID1304854
ABSTRAKT
The effect of 4-chlor-N-(cys-2,6-dimethyl peridine)-3-sulfomoin-benzamide as an anti-hypertensive drug on left-to-right hemisphere interactions was studied on experimental animals. Its effect on hemispheres was shown to be asymmetrical. The most effective action of the drug was demonstrated on the left hemisphere of rats differing in their trends of interhemispheric interactions.