- Preparation of 6-thioguanosine phosphoramidite for oligoribonucleotide synthesis.
Preparation of 6-thioguanosine phosphoramidite for oligoribonucleotide synthesis.
Nucleosides, nucleotides & nucleic acids (2003-10-21)
Yang Wang, Eric Lattmann, Qinguo Zheng
PMID14565391
ABSTRAKT
6-Thioguanosine phosphoramidite was prepared, using 2,4-dinitrophenyl as protection group for thio-function, and its stability towards conditions of RNA synthesis was investigated. The results show that the monomer was stable under the conditions of RNA synthesis and suitable for incorporation of thioguanine into oligoribonucleotides.