Merck
Neutrophil and platelet dynamics at organ level after cardiopulmonary bypass: an in vivo study in neonatal pigs.
APMIS : acta pathologica, microbiologica, et immunologica Scandinavica (2004-04-02)
V Brix-Christensen, M Rheling, C Flø, H Ravn, V Hjortdal, J Marqversen, N Andersen, E Tønnesen
ABSTRAKT
The aim was to investigate if organ dysfunction is a consequence of cell accumulation in the tissue and whether this accumulation is caused by the cardiopulmonary bypass (CPB) procedure. Twenty-six piglets were used in the sham group (sternotomy, n=12) or in the CPB group (sternotomy, CPB, n=14). Isotope-labeled autologous (99m)Tc-neutrophils (PMNs) and (111)In-platelets were infused and dynamically followed at organ level with a gamma camera before, during, and 4 h after termination of CPB. The CPB group showed a 49% increase in (99m) Tc-PMNs in the kidneys in the postoperative period compared to a decrease of 2% in the sham group. A less marked decrease was observed in the lungs and peripheral blood between the two groups. The increased radioactivity at organ level post-CPB could be due to changes in flow, extraction in the organ or accumulation of cells, especially in the kidneys and lungs, and might contribute to temporary organ dysfunction postoperatively.