Merck
  • Determination of a Metolazone metabolite in human urine by high-performance liquid chromatography/diode-array detection, high-performance liquid chromatography/electrospray ionization mass spectrometry and gas chromatography/mass spectrometry.
Determination of a Metolazone metabolite in human urine by high-performance liquid chromatography/diode-array detection, high-performance liquid chromatography/electrospray ionization mass spectrometry and gas chromatography/mass spectrometry.
Rapid communications in mass spectrometry : RCM (2004-04-20)
Yunje Kim, Sungil Kim, Myungsoo Kim, Won Lee
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Metolazone, ≥98% (HPLC), solid