Merck
Oligosaccharides implicated in recognition are predicted to have relatively ordered structures.
Biochemistry (2004-05-12)
Andrew Almond, Bent O Petersen, Jens Ø Duus
ABSTRAKT
Fucosylated O- and N-linked glycans are essential recognition molecules in plants and animals. To understand how they impart their functions, through interactions with proteins, requires a detailed analysis of structure and dynamics, but this is presently lacking. In this study, the three-dimensional structure and dynamics of three fucosylated oligosaccharides are investigated using a combination of high field (800 MHz) nuclear magnetic resonance and long (50 ns) molecular dynamics simulations in explicit water. Predictions from dynamics simulations were in agreement with nuclear Overhauser cross-peak intensities. Similarly, a theory of weak alignment in neutral media resulted in reasonable predictions of residual dipolar couplings for the trisaccharide fucosyllactose. However, for larger penta- and hexasaccharides (LNF-1 and LND-1), the anisotropic component of the alignment was underestimated, attributed to shape irregularities of the fucosyl branches on an otherwise linear core, being more pronounced in a singly branched than a doubly branched oligosaccharide. Simulations, confirmed by experiment, predicted fucosylated molecules that are restricted to librations about a single average conformation. This restriction is partly due to microscopic water interactions, which act to stabilize intramolecular hydrogen bonds and maintain tight and ordered conformations; a view not forthcoming from simpler, nonaqueous simulations. Such a conclusion is crucial for understanding how these molecules interact with proteins and impart their recognition properties.