Merck
Japanese pieris toxicosis of goats.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc (1992-07-01)
K H Plumlee, W G VanAlstine, J M Sullivan