- Alternating copolymerization of limonene oxide and carbon dioxide.
Alternating copolymerization of limonene oxide and carbon dioxide.
Journal of the American Chemical Society (2004-09-16)
Christopher M Byrne, Scott D Allen, Emil B Lobkovsky, Geoffrey W Coates
PMID15366863
ABSTRAKT
The alternating copolymerization of (R)- or (S)-limonene oxide and CO2 using beta-diiminate zinc acetate catalysts is reported. At 100 psi CO2 and 25 degrees C, the catalyst exhibits a high selectivity for the trans isomer and produces regioregular polycarbonate. The copolymer contains >99% carbonate linkages, a narrow molecular weight distribution, and an Mn value consistent with the [epoxide]/[Zn] ratio.