Merck
Experimental study on the antibacterial effect of origanum volatile oil on dysentery bacilli in vivo and in vitro.
Journal of Huazhong University of Science and Technology. Medical sciences = Hua zhong ke ji da xue xue bao. Yi xue Ying De wen ban = Huazhong keji daxue xuebao. Yixue Yingdewen ban (2004-12-14)
Fang Liao, Qinghua Huang, Zhende Yang, Hanlin Xu, Qinhua Gao
ABSTRAKT
To observe the germistatic and germicidal effects of origanum volatile oil (OVI) on the dysentery bacteria, the abdominal cavity of mice was infected with Shigella sonne (Sh. sonnei) and Shigella flexneri (Sh. flexneri) F2a. After OVI was given to the mice via gastric lavage, the effects of OVI on the infected mice were observed. The minimal inhibitory concentration (MIC) and minimal bactericidal concentration (MBC) for dysentery bacteria were determined in vitro. The results showed that origanum volatile oil showed obvious protective effect on mice infected with Sh. sonnei and Sh. flexneri F2a and it had germistatic and germicidal effects on dysentry bacteria. We are led to conclude that origanum volatile oil is an effective medicine against the infection of dysentery bacteria.