Merck
Infections of cefotaxime-resistant and cefmetazole-susceptible Escherichia coli and Klebsiella pneumoniae in children.
Journal of microbiology, immunology, and infection = Wei mian yu gan ran za zhi (2005-04-22)
Tsung Zu Wu, Po Ren Hsueh, Luan Yin Chang, Chun Yi Lu, Chung Yi Wang, Chin Ying Yang, Pei Lan Shao, Ping Ing Lee, Chin Yun Lee, Li Min Huang
ABSTRAKT
A search of the computerized database at the National Taiwan University Hospital was made for cefotaxime-resistant and cefmetazole-susceptible isolates of Escherichia coli and Klebsiella pneumoniae (which may be extended-spectrum beta-lactamase-producing strains) in pediatric wards and intensive care units between 1999 and 2001. Fourteen infectious episodes attributed only to study bacteria were identified, including 7 episodes of bacteremia. Nine patients (64.3%) had underlying medical conditions: 3 were premature babies, 3 were immunodeficient, 2 had malignancy, and 2 had a congenital heart disease with active heart failure even after surgery. Among the 7 patients with bacteremias, 5 may be catheter-related; 6 were treated with carbapenems and 1 was treated with cefmetazole successfully, with or without the removal of the catheter. Before the acquisition of the infection, a history of stay in an intensive care unit within 4 weeks was noted in 10 cases (71.4%); a history of use of extended-spectrum cephalosporins within 4 weeks was also noted in 6 cases (42.9%). Cefmetazole, with or without an aminoglycoside, was clinically effective in 6 cases (42.8%). Except for 1 episode of pneumonia that ended in mortality, all of the infectious episodes were successfully treated. The mortality rate was 7.1%.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Cefmetazole sodium salt