Merck
Alzheimer disease beta-amyloid activity mimics cholesterol oxidase.
The Journal of clinical investigation (2005-08-30)
Luigi Puglielli, Avi L Friedlich, Kenneth D R Setchell, Seiichi Nagano, Carlos Opazo, Robert A Cherny, Kevin J Barnham, John D Wade, Simon Melov, Dora M Kovacs, Ashley I Bush
ABSTRAKT
The abnormal accumulation of amyloid beta-peptide (Abeta) in the form of senile (or amyloid) plaques is one of the main characteristics of Alzheimer disease (AD). Both cholesterol and Cu2+ have been implicated in AD pathogenesis and plaque formation. Abeta binds Cu2+ with very high affinity, forming a redox-active complex that catalyzes H2O2 production from O2 and cholesterol. Here we show that Abeta:Cu2+ complexes oxidize cholesterol selectively at the C-3 hydroxyl group, catalytically producing 4-cholesten-3-one and therefore mimicking the activity of cholesterol oxidase, which is implicated in cardiovascular disease. Abeta toxicity in neuronal cultures correlated with this activity, which was inhibited by Cu2+ chelators including clioquinol. Cell death induced by staurosporine or H2O2 did not elevate 4-cholesten-3-one levels. Brain tissue from AD subjects had 98% more 4-cholesten-3-one than tissue from age-matched control subjects. We observed a similar increase in the brains of Tg2576 transgenic mice compared with nontransgenic littermates; the increase was inhibited by in vivo treatment with clioquinol, which suggests that brain Abeta accumulation elevates 4-cholesten-3-one levels in AD. Cu2+-mediated oxidation of cholesterol may be a pathogenic mechanism common to atherosclerosis and AD.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
(+)-4-Cholesten-3-one, 98%