Merck
Mechanisms underlying the inhibitory effects induced by pituitary adenylate cyclase-activating peptide in mouse ileum.
European journal of pharmacology (2005-09-28)
Maria Grazia Zizzo, Flavia Mulè, Rosa Serio
ABSTRAKT
The aim of this study was to investigate the signal transduction mechanisms underlying the inhibitory effect induced by pituitary adenylate cyclase activating peptide (PACAP-27) on the spontaneous contractile activity of longitudinal muscle of mouse ileum. Mechanical activity of ileal segments was recorded isometrically in vitro. PACAP-27 produced apamin-sensitive reduction of the amplitude of the spontaneous contractions. 9-(Tetrahydro-2-furanyl)-9H-purin-6-amine (SQ 22,536), adenylate cyclase inhibitor, or genistein and tyrphostin 25, tyrosine kinase inhibitors, had negligible effects on PACAP-27-induced inhibition. PACAP-27 effects were significantly inhibited by U-73122, phopholipase C (PLC) inhibitor, by 2-aminoethoxy-diphenylborate (2-APB), permeable blocker of inositol 1,4,5-triphosphate (IP3) receptors and by depletion of Ca2+ stores with cyclopiazonic acid or thapsigargin. Ryanodine did not reduce PACAP-27-inhibitory responses. We suggest that, in mouse ileum, the inhibitory responses to PACAP-27 involve stimulation of PLC, increased production of IP3 and localised Ca2+ release from intracellular stores, which could provide the opening of apamin-sensitive Ca2+-dependent K+ channels.