- Genotoxicity and transmission electron microscopy studies of molybdocene dichloride.
Genotoxicity and transmission electron microscopy studies of molybdocene dichloride.
Journal of inorganic biochemistry (2006-03-11)
Kate S Campbell, Amalanie J Foster, Carolyn T Dillon, Margaret M Harding
PMID16527357
ABSTRAKT
The cytotoxic effects of molybdocene dichloride (Cp(2)MoCl(2)) were investigated in V79 Chinese hamster lung cells using the micronucleus assay. Cp(2)MoCl(2) produced significant genotoxic damage whereby 0.2micronuclei/1000 binucleated cells were induced per muM of Cp(2)MoCl(2). Transmission electron microscopic analysis of thin-sectioned cells treated with Cp(2)MoCl(2) (300microM) showed distinct morphological alterations of the nuclei, condensation of chromatin, and a high incidence of polynucleated cells. Implications for the mechanism of antitumor action of molybdocene dichloride are discussed.