Merck
Sinorhizobium meliloti bluB is necessary for production of 5,6-dimethylbenzimidazole, the lower ligand of B12.
Proceedings of the National Academy of Sciences of the United States of America (2006-03-16)
Gordon R O Campbell, Michiko E Taga, Kavita Mistry, Javier Lloret, Peter J Anderson, John R Roth, Graham C Walker
ABSTRAKT
An insight into a previously unknown step in B(12) biosynthesis was unexpectedly obtained through our analysis of a mutant of the symbiotic nitrogen fixing bacterium Sinorhizobium meliloti. This mutant was identified based on its unusually bright fluorescence on plates containing the succinoglycan binding dye calcofluor. The mutant contains a Tn5 insertion in a gene that has not been characterized previously in S. meliloti. The closest known homolog is the bluB gene of Rhodobacter capsulatus, which is implicated in the biosynthesis of B(12) (cobalamin). The S. meliloti bluB mutant is unable to grow in minimal media and fails to establish a symbiosis with alfalfa, and these defects can be rescued by the addition of vitamin B(12) (cyanocobalamin) or the lower ligand of cobalamin, 5,6-dimethylbenzimidazole (DMB). Biochemical analysis demonstrated that the bluB mutant does not produce cobalamin unless DMB is supplied. Sequence comparison suggests that BluB is a member of the NADH/flavin mononucleotide (FMN)-dependent nitroreductase family, and we propose that it is involved in the conversion of FMN to DMB.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
5,6-Dimethylbenzimidazole, ≥99%