Merck
Synthesis of nonactin and the proposed structure of trilactone.
Organic letters (2006-06-16)
Yikang Wu, Ya-Ping Sun
ABSTRAKT
[reaction: see text] An efficient enantioselective route to nonactin using a novel beta-inversion of an Evans syn aldol to construct the THF ring is presented. Through total synthesis, the structure for trilactone proposed in the literature is shown likely to be incorrect.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Jonofor amonowy I, Selectophore, function tested, ≥98.0% ((Total homologs), HPLC)
Sigma-Aldrich
Nonactin, from Streptomyces griseus, ≥98.0% (Total homologs, HPLC)