- Quazepam and flurazepam: differential pharmacokinetic and pharmacodynamic characteristics.
Quazepam and flurazepam: differential pharmacokinetic and pharmacodynamic characteristics.
The Journal of clinical psychiatry (1991-09-01)
J M Hilbert, D Battista
PMID1680120
ABSTRAKT
Quazepam and flurazepam share pharmacokinetic properties that result in prevention of early-morning insomnia, daytime rebound anxiety, and withdrawal rebound insomnia. Yet sleep laboratory and performance studies demonstrated that during a 1- to 4-week administration period quazepam had a low potential for causing daytime drowsiness or impairment. This profile may be related to several factors, such as differences in quazepam's metabolic pathways; plasma pharmacokinetics; rate of brain uptake, redistribution, and clearance; as well as differences in receptor binding and kinetics.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Desalkylflurazepam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®