Merck
[Psoriasis and anxiety: marked improvement after anxiolytic therapy].
Anales de pediatria (Barcelona, Spain : 2003) (2006-09-08)
A Figueroa Quintana, A Díez Suárez, A España Alonso, C Soutullo Esperón