Merck
Determination of the analgesic components of Spasmomigraine tablet by liquid chromatography with ultraviolet detection.
Journal of AOAC International (2007-03-22)
Fawzy A Elbarbry, Mokhtar M Mabrouk, Mohamed A El-Dawy
ABSTRAKT
A procedure was developed for the determination of the analgesic components of Spasmomigraine tablets, which are ergotamine (I), propyphenazone (II), caffeine (III), camylofin (IV), and mecloxamine (V). They were subjected to high-performance liquid chromatography on a column (300 x 3.9 mm, 10 rlm particle size) packed with micro-Bondapak C18. Separations were achieved with the mobile phase methanol-water-triethylamine (60 + 40 + 0.1, v/v/v) flowing at a rate of 1.5 mL/min, and quantitative determination was performed at 254 nm at ambient temperature for I-III; acetonitrile-25 mM KH2PO4-acetic acid (45 + 55 + 0.2, v/v/v), flowing at a rate of 1.5 mL/min and detection at 234 nm at ambient temperature, was used for IV and V. Methyl paraben was used as an internal standard. The detection limits were 0.35 (I), 5.0 (11), 1.5 (111), 3.0 (IV), and 2.0 microg/mL (V). The method was accurate (mean recovery 98+/-2%, n = 4) and precise (coefficient of variation <5%, n = 5). The proposed method is rapid and sensitive and, therefore, suitable for the routine control of these ingredients in multicomponent dosage forms.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Propyphenazone, European Pharmacopoeia (EP) Reference Standard
Supelco
Propyphenazone, Pharmaceutical Secondary Standard; Certified Reference Material