Merck
Antidepressant-like effects of Albizzia julibrissin in mice: involvement of the 5-HT1A receptor system.
Pharmacology, biochemistry, and behavior (2007-05-05)
Ji-Hyun Kim, Sun Yeou Kim, Seok-Yong Lee, Choon-Gon Jang
ABSTRAKT
The present study was undertaken to investigate the antidepressant-like effects of the methylene chloride fraction of Albizzia julibrissin (MCAJ) using a tail suspension test in mice. MCAJ was orally administered at 50, 100, or 200 mg/kg to mice, 1 h before the tail suspension test. Acute treatment with MCAJ at 200 mg/kg significantly reduced the immobility time compared with the control group, and thus showed an antidepressant-like effect. This effect was comparable to that of imipramine at 10 mg/kg. This antidepressant-like effect was reversed by treatment with WAY-100635 (a 5-HT1A receptor antagonist) or pindolol (a 5-HT1A/1B receptor antagonist). However, the antidepressant effect of MCAJ was not effected by treatment with GR55562 (a 5-HT1B receptor antagonist) or ketanserin (a 5-HT2A receptor antagonist). Therefore, our findings suggest that MCAJ exerts its antidepressant-like effect via the 5-HT1A receptor system.