Merck
  • Stereoselective hexobarbital 3'-hydroxylation by CYP2C19 expressed in yeast cells and the roles of amino acid residues at positions 300 and 476.
Stereoselective hexobarbital 3'-hydroxylation by CYP2C19 expressed in yeast cells and the roles of amino acid residues at positions 300 and 476.
Chirality (2007-05-10)
Keita Saito, Hikari Dan, Kazufumi Masuda, Takashi Katsu, Nobumitsu Hanioka, Shigeo Yamamoto, Kazuko Miyano, Shigeru Yamano, Shizuo Narimatsu
ABSTRAKT
We examined the enzymatic function of recombinant CYP2C19 in enantiomeric hexobarbital (HB) 3'-hydroxylation, and searched the roles of amino acid residues, such as Phe-100, Phe-114, Asp-293, Glu-300, and Phe-476 of CYP2C19 in the stereoselective HB 3'-hydroxylation, using a yeast cell expression system and site-directed mutagenesis method. CYP2C19 wild-type exerted substrate enantioselectivity of (R)-HB>(S)-HB and metabolite diastereoselectivity of 3'(R)<3'(S) in 3'-hydroxylation of HB enantiomers. The substitution of Asp-293 by alanine failed to yield an observable peak at 450 nm in its reduced carbon monoxide-difference spectrum. CYP2C19-E300A and CYP2C19-E300V with alanine and valine, respectively, in place of Glu-300 exerted total HB 3'-hydroxylation activities of 45 and 108%, respectively, that of the wild-type. Interestingly, these two mutants showed substrate enantioselectivity of (R)-HB<(S)-HB, which is opposite to that of the wild-type, while metabolite diasteroselectivity remained unchanged. The replacement of Phe-476 by alanine increased total HB 3'-hydroxylation activity to approximately 3-fold that of the wild-type. Particularly, 3'(S)-OH-(S)-HB-forming activity elevated to 7-fold that of the wild-type, resulting in the reversal of the substrate enantioselectivity. In contrast, the substitution of phenylalanine at positions 100 and 114 by alanine did not produce a remarkable change in the total activity or the substrate enantioselectivity. These results indicate that Glu-300 and Phe-476 are important in stereoselective oxidation of HB enantiomers by CYP2C19.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Hexobarbital solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®