Merck
  • Elucidation of urinary metabolites of fluoxymesterone by liquid chromatography-tandem mass spectrometry and gas chromatography-mass spectrometry.
Elucidation of urinary metabolites of fluoxymesterone by liquid chromatography-tandem mass spectrometry and gas chromatography-mass spectrometry.
Journal of mass spectrometry : JMS (2007-11-27)
Oscar J Pozo, Wim Van Thuyne, Koen Deventer, Peter Van Eenoo, Frans T Delbeke
ABSTRAKT
The suitability of liquid chromatography tandem mass spectrometry (LC-MS/MS) and gas chromatography mass spectrometry (GC-MS) for the elucidation of fluoxymesterone metabolism has been evaluated. Electrospray ionization (ESI) and collision induced dissociation (CID) fragmentation in LC-MS/MS and electron impact spectra (EI) in GC-MS have been studied for fluoxymesterone and two commercially available metabolites. MS(n) experiments and accurate mass measurements performed by an ion-trap analyser and a QTOF instrument respectively have been used for the elucidation of the fragmentation pathway. The neutral loss scan of 20 Da (loss of HF) in LC-MS/MS has been applied for the selective detection of fluoxymesterone metabolites. In a positive fluoxymesterone doping control sample, 9 different analytes have been detected including the parent compound. Seven of these metabolites were also confirmed by GC-MS including 5 previously unreported metabolites. On the basis of the ionization, the CID fragmentation, the accurate mass of the product ions and the EI spectra of these analytes, a tentative elucidation as well as a proposal for the metabolic pathway of fluoxymesterone has been suggested. The presence of these compounds has also been confirmed by the analysis of five other positive fluoxymesterone urine samples.