Merck
Determination of active enzyme concentration using activity-based probes and direct mass spectrometric readout.
Analytical biochemistry (2007-12-11)
Cédric Bovet, Renato Zenobi
ABSTRAKT
Activity-based probes (ABPs) are specific covalent inhibitors developed for different classes of enzymes. We have titrated a serine protease and a lipase with their specific ABPs and measured the extent of inhibition using nanoelectrospray mass spectrometry (nanoESI-MS). Because ABPs only interact with the active enzyme form, the approach allows to accurately measure the active enzyme concentration in solution. This is even possible in the presence of contaminants. The concentrations of the two enzymes were also investigated by UV spectroscopy, which appears to give higher concentrations than those measured with the active site titration method.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
p-Toluenesulfonyl fluoride, 98%