Merck
Effects of oral and transdermal hormonal contraception on vascular risk markers: a randomized controlled trial.
Obstetrics and gynecology (2008-02-02)
Julia V Johnson, Jane Lowell, Gary J Badger, Jan Rosing, Svetlana Tchaikovski, Mary Cushman
ABSTRAKT
To compare the effects of oral and transdermal contraceptives containing similar hormone formulations on vascular risk markers. We conducted a randomized, investigator-blinded, crossover, clinical trial with 24 healthy women, aged 18-35 years, who received 2 months of transdermal or oral contraceptive, 2 months washout, then 2 months of the alternative medication. The transdermal contraceptive contained 0.75 mg ethinyl estradiol and 6 mg norelgestromin. The oral contraceptive contained 35 mcg ethinyl estradiol and 250 mcg norgestimate. Blood samples taken before and after each treatment were analyzed in batch for D-dimer, von Willebrand factor, factor VIII, total and free protein S, antithrombin, fibrinogen, C-reactive protein, and normalized activated protein C sensitivity ratio (nAPCsr) determined with two thrombin generation-based assays, the alpha2macroglobulin-thrombin end point method (alpha2M-IIa) and calibrated automated thrombinography. Repeated measures analysis of variance was used for analysis. For both contraceptives (transdermal, oral) there were significant declines in free (19%, 11%) and total protein S (19%, 13%) and antithrombin (13%, 10%); increases in fibrinogen (8%, 10%), C-reactive protein (220%, 292%), nAPCsr alpha2M-IIa (81%, 61%), and nAPCsr calibrated automated thrombinography (102%, 68%), all P<.05. Transdermal contraceptives had a greater effect than oral contraceptives on free protein S (P=.07), nAPCsr alpha2M-IIa (P=.06), and nAPCsr calibrated automated thrombinography (P=.03). Oral and transdermal contraception with similar hormones had similar adverse effects on vascular risk markers. This suggests that this transdermal contraceptive has at least a similar thrombosis risk as its oral counterpart. ClinicalTrials.gov, www.clinicaltrials.gov, NCT00554632 I.