Merck
Total synthesis of (+/-)-actinophyllic acid.
Journal of the American Chemical Society (2008-05-22)
Connor L Martin, Larry E Overman, Jason M Rohde
ABSTRAKT
The first total synthesis of (+/-)-actinophyllic acid (1) is reported. Key steps of this synthesis include an intramolecular oxidative coupling of ketone and malonic ester enolates and an aza-Cope-Mannich rearrangement that assembled the core structure of the natural product's unique ring system. The synthesis was accomplished from di-tert-butyl malonate in 8% overall yield by a concise sequence that proceeds by way of only seven isolated intermediates.