Merck
Identification of gangliosides recognized by IgG anti-GalNAc-GD1a antibodies in bovine spinal motor neurons and motor nerves.
Brain research (2008-07-05)
Hiide Yoshino, Toshio Ariga, Akemi Suzuki, Robert K Yu, Tadashi Miyatake
ABSTRAKT
The presence of immunoglobulin G (IgG)-type antibodies to the ganglioside, N-acetylgalactosaminyl GD1a (GalNAc-GD1a), is closely associated with the pure motor type of Guillain-Barré syndrome (GBS). In the present study, we isolated disialogangliosides from the motor neurons and motor nerves of bovine spinal cords by DEAE-Sephadex column chromatography. The disialoganglioside fraction contained GD1a, GD2, GD1b, and three gangliosides, designated X1, X2 and X3. Serum from a patient with axonal GBS with IgG anti-GalNAc-GD1a antibody yielded positive immunostaining with X1, X2, and X3. When isolated by preparative thin-layer chromatography (TLC), X1 migrated at the same position as GalNAc-GD1a from Tay-Sachs brain, suggesting that X1 is GalNAc-GD1a containing N-acetylneuraminic acid (NeuAc). TLC of isolated X2 revealed that it migrated between GD1a and GD2. On the other hand, X3 had a migratory rate on TLC between and GD1b and GT1b. Since both X2 and X3 were recognized by IgG anti-GalNAc-GD1a antibody, the results suggest that X2 is a GalNAc-GD1a species containing a mixture containing a NeuAc-and an N-glycolylneuraminic acid (NeuGc) species, and X3 is a GalNAc-GD1a species with two NeuGc. This evidence indicating the specific localization of GalNAc-GD1a and its isomers in spinal motor neurons should be useful in elucidating the pathogenic role of IgG anti-GalNAc-GD1a antibody in pure motor-type GBS.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Disialoganglioside GD1b from bovine brain, ~95%, lyophilized powder