Merck
Beta-cell alpha-ketoglutarate hydroxylases may acutely participate in insulin secretion.
Metabolism: clinical and experimental (2008-07-22)
Michael J Fallon, Michael J MacDonald
ABSTRAKT
The presence of Fe(II) alpha-ketoglutarate hydroxylases in rat and human pancreatic islets and INS-1 832/13 cells was demonstrated with the reverse transcriptase polymerase chain reaction (PHD1, 2, and 3; lysyl hydroxylases 1, 2, and 3; and phytanoyl-coenzyme A hydroxylase were seen) and/or immunoblotting (high levels of proline hydroxylase P4Halpha1, PHD2, and PHD4 and low levels of PHD2 and PHD3 in human islets, and high levels of PHD2 in rat islets and INS-1 cells were seen). Prolyl hydroxylase enzyme activity in INS-1 832/13 cells was purified with polyproline affinity chromatography. Inhibitors of alpha-ketoglutarate hydroxylases lowered glucose-induced and leucine-plus-glutamine-induced insulin release in rat pancreatic islets, suggesting that there may be acute unknown effects of alpha-ketoglutarate hydroxylases in insulin secretion. It is possible that an increase in mitochondrially generated alpha-ketoglutarate derived from insulin secretagogue carbon and translocated to the cytosol may be part of the signal for insulin secretion.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Protocatechuic acid ethyl ester, 97%