- Four-component benzyne coupling reactions: a concise total synthesis of dehydroaltenuene B.
Four-component benzyne coupling reactions: a concise total synthesis of dehydroaltenuene B.
Organic letters (2008-08-05)
Darunee Soorukram, Tao Qu, Anthony G M Barrett
PMID18672878
ABSTRAKT
A four-component coupling reaction of 3,5-dimethoxybenzyne with 2-methyl-2-cyclohexenylmagnesium chloride, carbon dioxide, and iodine was utilized as a key step in the first total synthesis of dehydroaltenuene B.