Merck
  • Experimental and molecular dynamics simulation study of the sublimation energetics of cyclopentadienyltricarbonylmanganese (Cymantrene).
Experimental and molecular dynamics simulation study of the sublimation energetics of cyclopentadienyltricarbonylmanganese (Cymantrene).
The journal of physical chemistry. A (2008-09-20)
Ricardo Picciochi, José N Canongia Lopes, Hermínio P Diogo, Manuel E Minas da Piedade
ABSTRAKT
The standard molar enthalpy of sublimation of monoclinic cyclopentadienyltricarbonylmanganese, Mn(eta (5)-C 5H 5)(CO) 3, at 298.15 K, was determined as Delta sub H m (o)[Mn(eta (5)-C 5H 5)(CO) 3] = 75.97 +/- 0.37 kJ x mol (-1) from Knudsen effusion and Calvet-drop microcalorimetry measurements, thus considerably improving the very large inaccuracy (>10 kJ x mol (-1)) of the published data. The obtained value was used to assess the extension of the OPLS-based all-atom force field we previously developed for iron metallocenes to manganese organometallic compounds. The modified force field was able to reproduce the volumetric properties (density and unit-cell volume) of crystalline Mn(eta (5)-C 5H 5)(CO) 3 with a deviation of 0.6% and the experimentally determined enthalpy of sublimation with an accuracy of 1 kJ x mol (-1). The interaction (epsilon) and atomic-diameter (sigma) parameters of the Lennard-Jones (12-6) potential function used to calculate dispersion contributions within the framework of the force field were found to be transferable from iron to manganese.