Merck
Using directed evolution to probe the substrate specificity of mandelamide hydrolase.
Protein engineering, design & selection : PEDS (2008-12-17)
Pan-Fen Wang, Alejandra Yep, George L Kenyon, Michael J McLeish
ABSTRAKT
Mandelamide hydrolase (MAH), a member of the amidase signature family, catalyzes the hydrolysis of mandelamide to mandelate and ammonia. X-ray structures of several members of this family, but not that of MAH, have been reported. These reveal nearly superimposable conformations of the unusual Ser-cisSer-Lys catalytic triad. Conversely, the residues involved in substrate recognition are not conserved, implying that the binding pocket could be modified to change the substrate specificity, perhaps by directed evolution. Here we show that MAH is able to hydrolyze small aliphatic substrates such as lactamide, albeit with low efficiency. A selection method to monitor changes in mandelamide/lactamide preference was developed and used to identify several mutations affecting substrate binding. A homology model places some of these mutations close to the catalytic triad, presumably in the MAH active site. In particular, Gly202 appears to control the preference for aromatic substrates as the G202A variant showed three orders of magnitude decrease in k(cat)/K(m) for (R)- and (S)-mandelamide. This reduction in activity increased to six orders of magnitude for the G202V variant.