Merck
Investigation of the effects of the novel anticonvulsant compound carisbamate (RWJ-333369) on rat piriform cortical neurones in vitro.
British journal of pharmacology (2009-02-20)
B J Whalley, G J Stephens, A Constanti
ABSTRAKT
Carisbamate is being developed for adjuvant treatment of partial onset epilepsy. Carisbamate produces anticonvulsant effects in primary generalized, complex partial and absence-type seizure models, and exhibits neuroprotective and antiepileptogenic properties in rodent epilepsy models. Phase IIb clinical trials of carisbamate demonstrated efficacy against partial onset seizures; however, its mechanisms of action remain unknown. Here, we report the effects of carisbamate on membrane properties, evoked and spontaneous synaptic transmission and induced epileptiform discharges in layer II-III neurones in piriform cortical brain slices. Effects of carisbamate were investigated in rat piriform cortical neurones by using intracellular electrophysiological recordings. Carisbamate (50-400 micromol x L(-1)) reversibly decreased amplitude, duration and rise-time of evoked action potentials and inhibited repetitive firing, consistent with use-dependent Na+ channel block; 150-400 micromol x L(-1) carisbamate reduced neuronal input resistance, without altering membrane potential. After microelectrode intracellular Cl(-) loading, carisbamate depolarized cells, an effect reversed by picrotoxin. Carisbamate (100-400 micromol x L(-1)) also selectively depressed lateral olfactory tract-afferent evoked excitatory synaptic transmission (opposed by picrotoxin), consistent with activation of a presynaptic Cl(-) conductance. Lidocaine (40-320 micromol x L(-1)) mimicked carisbamate, implying similar modes of action. Carisbamate (300-600 micromol x L(-1)) had no effect on spontaneous GABA(A) miniature inhibitory postsynaptic currents and at lower concentrations (50-200 micromol x L(-1)) inhibited Mg2+-free or 4-aminopyridine-induced seizure-like discharges. Carisbamate blocked evoked action potentials use-dependently, consistent with a primary action on Na+ channels and increased Cl(-) conductances presynaptically and, under certain conditions, postsynaptically to selectively depress excitatory neurotransmission in piriform cortical layer Ia-afferent terminals.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
4-Aminopyrimidine, 98%