- Glycogen phosphorylase a inhibitors with a phenethylphenylphthalimide skeleton derived from thalidomide-related alpha-glucosidase inhibitors and liver X receptor antagonists.
Glycogen phosphorylase a inhibitors with a phenethylphenylphthalimide skeleton derived from thalidomide-related alpha-glucosidase inhibitors and liver X receptor antagonists.
Biological & pharmaceutical bulletin (2009-09-02)
Kazunori Motoshima, Minoru Ishikawa, Kazuyuki Sugita, Yuichi Hashimoto
PMID19721243
ABSTRAKT
Novel glycogen phosphorylase a (GPa) inhibitors with a phenethylphenylphthalimide skeleton were prepared based on alpha-glucosidase inhibitors and liver X receptor (LXR) antagonists derived from thalidomide. Their structure-activity relationships were analyzed. Some of the compounds thus prepared showed potent inhibitory activity against rabbit muscle GPa with more than 10-fold greater efficacy than a typical GPa inhibitor, 1,4-dideoxy-1,4-imino-D-arabinitol.