Merck
Absence of adenosine-mediated aortic relaxation in A(2A) adenosine receptor knockout mice.
American journal of physiology. Heart and circulatory physiology (2009-09-15)
Dovenia S Ponnoth, Maryam Sharifi Sanjani, Catherine Ledent, Kevin Roush, Thomas Krahn, S Jamal Mustafa
ABSTRAKT
Adenosine mediates vascular responses through four receptor subtypes: A(1), A(2A), A(2B), and A(3). The role of A(2A) receptors in aortic vascular tone was investigated using A(2A) adenosine receptor (AR) knockout (A(2A)KO) and corresponding wild-type (A(2A)WT) mice. Isolated aortic rings from A(2A)WT and A(2A)KO mice were precontracted with phenylephrine (10(-7) M), and concentration responses for adenosine analogs and selective agonists/antagonists were obtained. Nonselective adenosine analog (NECA; EC(50) = 6.78 microM) and CGS-21680 (A(2A)AR selective agonist; EC(50) = 0.013 microM) produced concentration-dependent relaxation (maximum of 25% and 28% relaxation at 10(-5) M NECA and CGS-21680, respectively) in A(2A)WT aorta. In A(2A)KO aorta, NECA (EC(50) = 0.075 microM) induced concentration-dependent contraction (maximum contraction of 47% at 10(-6) M; P < 0.05 compared with A(2A)WT), whereas CGS-21680 produced no response. SCH-58261 (10(-6) M; A(2A)AR selective antagonist) abolished both NECA- and CGS-21680-mediated vasorelaxation in A(2A)WT (P < 0.05), whereas no change was observed in A(2A)KO. When DPCPX (10(-5) M; A(1) selective antagonist) was used in NECA concentration response, greater vasorelaxation was observed in A(2A)WT (50% vs. 25% in controls at 10(-5) M; P < 0.05), whereas lower contraction was seen in A(2A)KO tissues (5% vs. 47% in controls at 10(-6) M; P < 0.05). Aortic endothelial function, determined by response to acetylcholine, was significantly higher in WT compared with KO (66% vs. 51%; P < 0.05). BAY 60-6583 (A(2B) selective agonist) produced similar relaxation in both KO and WT tissues. In conclusion, A(2A)AR KO mice had significantly lower aortic relaxation and endothelial function, suggesting that the A(2A)AR plays an important role in vasorelaxation, probably through an endothelium-dependent mechanism.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Alloxazine, 96%