Merck
Stanozolol treatment for successful prevention of attacks of severe primary cryofibrinogenemia.
Pediatric blood & cancer (2010-03-09)
Sule Unal, Fehime Kara, Seza Ozen, Diclehan Orhan, Murat Tuncer, Fatma Gumruk
ABSTRAKT
Severe essential cryofibrinogenemia is rare in childhood, and both the diagnosis and the management are challenging for pediatricians. An 11-year-old male, who had already lost two digits following cold exposure, was referred after multiple visits to various hospitals and subsequently diagnosed as primary cryofibrinogenemia. His history revealed unresponsiveness to calcium channel blockers, acetyl salicylic acid, pentoxifylline, dextran, and steroids. Stanozolol (2 mg/day, orally) prophylaxis was initiated and no new skin lesions developed following starting this treatment. Some of the newly formed lesions at the onset of stanozolol healed.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Stanozolol solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®