Merck
Detection of mono-hydroxylated metabolites of stanozolol by HPLC-ESI (+) MS/MS in Indian sports persons.
Drug testing and analysis (2010-04-01)
S Ahi, A Beotra, S Jain
ABSTRAKT
The abuse of stanozolol is quite widespread in Indian sport. Its analysis is challenging and this has led to the development of new methods to improve its detection. A method was developed and validated for the detection of the three main monohydroxylated metabolites of stanozolol. The excretion profile of these metabolites was studied in four healthy male volunteers. The excretion study samples, after a single oral dose of drug, showed that 3'-OH-stanozolol was excreted at the highest concentration, followed by 16beta-OH stanozolol, with 4beta-OH stanozolol as the least excreted. Ninety-eight old doping samples with adverse analytical findings for 3'-OH-stanozolol were reanalysed using this method. This showed 3'-OH-stanozolol and 16beta-OH stanozolol in all the 98 samples whereas 4beta-OH-stanozolol was identified in 90 samples. The percentage of positive identifications of stanozolol in Indian sportspeople has increased markedly in the last five years, from 31.9% in 2004 to 81.8% in 2009; however, this may be due to the more effective detection of stanozolol metabolites. It can thus be concluded that the marked increase in percent positive of stanozolol in Indian sportspersons in 2009 may be due to the improved detection by a more effective LCMS/MS method.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Supelco
Stanozolol solution, 1.0 mg/mL in 1,2-dimethoxyethane, ampule of 1 mL, certified reference material, Cerilliant®