Merck
  • Interactions of nicotinamide-adenine dinucleotide phosphate analogues and fragments with pigeon liver malic enzyme. Synergistic effect between the nicotinamide and adenine moieties.
Interactions of nicotinamide-adenine dinucleotide phosphate analogues and fragments with pigeon liver malic enzyme. Synergistic effect between the nicotinamide and adenine moieties.
The Biochemical journal (1987-07-15)
H J Lee, G G Chang
ABSTRAKT
The structural requirements of the NADP+ molecule as a coenzyme in the oxidative decarboxylation reaction catalysed by pigeon liver malic enzyme were studied by kinetic and fluorimetric analyses with various NADP+ analogues and fragments. The substrate L-malate had little effect on the nucleotide binding. Etheno-NADP+, 3-acetylpyridine-adenine dinucleotide phosphate, and nicotinamide-hypoxanthine dinucleotide phosphate act as alternative coenzymes for the enzyme. Their kinetic parameters were similar to that of NADP+. Thionicotinamide-adenine dinucleotide phosphate, 3-aminopyridine-adenine dinucleotide phosphate, 5'-adenylyl imidodiphosphate, nicotinamide-adenine dinucleotide 3'-phosphate and NAD+ act as inhibitors for the enzyme. The first two were competitive with respect to NADP+ and non-competitive with respect to L-malate; the other inhibitors were non-competitive with NADP+. All NADP+ fragments were inhibitory to the enzyme, with a wide range of affinity, depending on the presence or absence of a 2'-phosphate group. Compounds with this group bind to the enzyme 2-3 orders of magnitude more tightly than those without this group. Only compounds with this group were competitive inhibitors with respect to NADP+. We conclude that the 2'-phosphate group is crucial for the nucleotide binding of this enzyme, whereas the carboxyamide carbonyl group of the nicotinamide moiety is important for the coenzyme activity. There is a strong synergistic effect between the binding of the nicotinamide and adenosine moieties of the nucleotide molecule.