Merck
  • The acyl-coenzyme A: cholesterol acyltransferase inhibitor CI-1011 reverses diffuse brain amyloid pathology in aged amyloid precursor protein transgenic mice.
The acyl-coenzyme A: cholesterol acyltransferase inhibitor CI-1011 reverses diffuse brain amyloid pathology in aged amyloid precursor protein transgenic mice.
Journal of neuropathology and experimental neurology (2010-07-09)
Henri J Huttunen, Daniel Havas, Camilla Peach, Cory Barren, Stephan Duller, Weiming Xia, Matthew P Frosch, Birgit Hutter-Paier, Manfred Windisch, Dora M Kovacs
ABSTRAKT
Cerebral accumulation of amyloid-beta (Abeta) is characteristic of Alzheimer disease and of amyloid precursor protein (APP) transgenic mice. Here, we assessed the efficacy of CI-1011, an inhibitor of acyl-coenzyme A:cholesterol acyltransferase, which is suitable for clinical use, in reducing amyloid pathology in both young (6.5 months old) and aged (16 months old) human APP transgenic mice. Treatment of young animals with CI-1011 decreased amyloid plaque load in the cortex and hippocampus and reduced the levels of insoluble Abeta40 and Abeta42 and C-terminal fragments of APP in brain extracts. In aged mice, CI-1011 specifically reduced diffuse amyloid plaques with a minor effect on thioflavin S-positive dense-core plaques. Reduced diffusible amyloid was accompanied by suppression of astrogliosis and enhanced microglial activation. Collectively, these data suggest that CI-1011 treatment reduces amyloid burden in human APP mice by limiting generation and increasing clearance of diffusible Abeta.
MATERIAŁY
Numer produktu
Marka
Opis produktu
Sigma-Aldrich
Avasimibe, ≥98% (HPLC)
Sigma-Aldrich
Anti-Amyloid Precursor Protein, C-Terminal antibody produced in rabbit, IgG fraction of antiserum, buffered aqueous solution